missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ro 60-0175 fumarate

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent, selective 5-HT2 receptor agonist; shows selectivity for the 5-HT2C subtype (pKi values are 9, 7.5, 5.4, 5.2 and 5.6 for human 5-HT2C, 2A, 1A, 6 and 7 receptors respectively). Centrally active following oral or systemic administration in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 342.75 |
| Purity | >98% |
| Molecular Weight (g/mol) | 342.75 |
| Chemical Name or Material | Ro 60-0175 fumarate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction