missing translation for 'onlineSavingsMsg'
Learn More

Robenidine hydrochloride, MedChemExpress

Product Code. 16448856
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448856 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16448856 Supplier Medchem Express Supplier No. HYB2157
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Robenidine hydrochloride is an anticoccidial agent which is also active against MRSA and VRE with MIC50s of 8.1 and 4.7 μM, respectively.

Robenidine hydrochloride is an anticoccidial agent which is also active against MRSA and VRE with MIC50s of 8.1 and 4.7 μM, respectively.

Specifications

Chemical Name or Material Robenidine hydrochloride
Physical Form Solid
CAS 25875-50-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C15H14Cl3N5
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 6.25 mg/mL (16.86 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES N=C(N/N=C/C1=CC=C(Cl)C=C1)N/N=C/C2=CC=C(Cl)C=C2.[H]Cl
Molecular Weight (g/mol) 370.66
Formula Weight 370.66
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.