missing translation for 'onlineSavingsMsg'
Learn More

ROCK-IN-2, MedChemExpress

Product Code. 16486527
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486527 5 mg 5mg
16454856 100 mg 100mg
16423808 10 mg 10mg
16410187 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16486527 Supplier Medchem Express Supplier No. HY10319
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress ROCK-IN-2 (Azaindole 1; TC-S 7001) is an orally active and ATP-competitive ROCK inhibitor with IC50s of 0.6 and 1.1 nM for human ROCK-1 and ROCK-2, respectively.

ROCK-IN-2 (Azaindole 1

Specifications

Chemical Name or Material ROCK-IN-2
CAS 867017-68-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H13ClF2N6O
Quantity 5 mg
Synonym Azaindole 1 TC-S 7001
Solubility Information DMSO : 33.33 mg/mL (82.75 mM; Need ultrasonic)
SMILES NC1=NC(Cl)=CC(NC2=CC(F)=C(OC3=C4C(NC=C4C)=NC=C3)C(F)=C2)=N1
Molecular Weight (g/mol) 402.79
Formula Weight 402.79
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.