missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROCK2 Antibody (S01-4F1), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00 € - 645.00 €
Specifications
| Antigen | ROCK2 |
|---|---|
| Clone | S01-4F1 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot, ELISA, Immunofluorescence, Sandwich ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18657054
|
Novus Biologicals
NBP3-19508-100ul |
100 μg |
645.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18667064
|
Novus Biologicals
NBP3-19508-50ul |
50 μg |
421.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ROCK2 Monoclonal antibody specifically detects ROCK2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISASpecifications
| ROCK2 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| coiled-coil-containing protein kinase 2, EC 2.7.11, EC 2.7.11.1, KIAA0619, p164 ROCK-2, Rho kinase 2, rho-associated protein kinase 2, Rho-associated, coiled-coil containing protein kinase 2 | |
| A synthetic peptide of human ROCK2 (Uniprot # O75116) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S01-4F1 | |
| Western Blot, ELISA, Immunofluorescence, Sandwich ELISA | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Phospho Specific, Protein Kinase, Wnt Signaling Pathway | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 9475 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title