missing translation for 'onlineSavingsMsg'
Learn More

Rolitetracycline, MedChemExpress

Product Code. 16489967
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489967 5 mg 5mg
16467488 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16489967 Supplier Medchem Express Supplier No. HY18257
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Rolitetracycline, a derivative of tetracycline, is a broad-spectrum antibiotic. Rolitetracyclin has a role as a protein synthesis inhibitor, an antiprotozoal drug and a prodrug.

Rolitetracycline, a derivative of tetracycline, is a broad-spectrum antibiotic. Rolitetracyclin has a role as a protein synthesis inhibitor, an antiprotozoal drug and a prodrug.

Specifications

Chemical Name or Material Rolitetracycline
CAS 751-97-3
Color Light Brown
Purity Grade Notes Research
Molecular Formula C27H33N3O8
Quantity 5 mg
Solubility Information H2O : 10 mg/mL (18.95 mM; Need ultrasonic and warming) ∣Ethanol : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble)
SMILES O=C(C(C1=O)=C(O)[C@@H](N(C)C)[C@]2([H])C[C@]3([H])[C@](C)(O)C4=C(C(C3=C(O)[C@@]21O)=O)C(O)=CC=C4)NCN5CCCC5
Molecular Weight (g/mol) 527.57
Formula Weight 527.57
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.