missing translation for 'onlineSavingsMsg'
Learn More

Ronidazole, MedChemExpress

Product Code. 16422239
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422239 1 g 1g
16490998 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16422239 Supplier Medchem Express Supplier No. HYB0565
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ronidazole is a potent and orally active antiprotozoal and anti-microbial agent. Ronidazole acts as a veterinary agent against Tritrichomonas foetus in cats models. Ronidazole can be used the research of forhistomoniasis and swine dysentery.

Ronidazole is a potent and orally active antiprotozoal and anti-microbial agent. Ronidazole acts as a veterinary agent against Tritrichomonas foetus in cats models. Ronidazole can be used the research of forhistomoniasis and swine dysentery.

Specifications

Chemical Name or Material Ronidazole
Physical Form Powder
CAS 7681-76-7
Color Yellow
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C6H8N4O4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (499.63 mM; Need ultrasonic) ∣H2O : 4 mg/mL (19.99 mM; Need ultrasonic)
SMILES NC(OCC1=NC=C([N+]([O-])=O)N1C)=O
Molecular Weight (g/mol) 200.15
Formula Weight 200.15
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.