missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ropporin 1-like Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99172-100ul
This item is not returnable.
View return policy
Description
Ropporin 1-like Polyclonal antibody specifically detects Ropporin 1-like in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Ropporin 1-like | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| AKAP-associated sperm protein, ASPFLJ23003, FLJ25776, radial spoke head 11 homolog, rhophilin associated tail protein 1-like, ROPN1-like protein, ropporin 1-like, ropporin-1-like protein, RSPH11 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Ropporin 1-like. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 83853 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction