missing translation for 'onlineSavingsMsg'
Learn More

Roquinimex, MedChemExpress

Product Code. 16441987
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16441987

Brand: Medchem Express HY13743

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Roquinimex (Linomide; PNU212616; ABR212616) is a quinoline derivative immunostimulant which increases NK cell activity and macrophage cytotoxicity; inhibits angiogenesis and reduces the secretion of TNF alpha.

Roquinimex (Linomide

Specifications

Chemical Name or Material Roquinimex
Physical Form Solid
CAS 84088-42-6
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H16N2O3
For Use With (Application) COVID-19-immunoregulation
Synonym Linomide FCF89 ABR212616
Solubility Information DMSO : ≥ 83.3 mg/mL (270.17 mM)
SMILES O=C(C1=C(O)C2=C(N(C)C1=O)C=CC=C2)N(C)C3=CC=CC=C3
Molecular Weight (g/mol) 308.33
Formula Weight 308.33
Percent Purity 98.88%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.