missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR beta Antibody (SR1948), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22118-100ul
This item is not returnable.
View return policy
Description
ROR beta Monoclonal antibody specifically detects ROR beta in Human, Mouse samples. It is validated for Western Blot
Specifications
| ROR beta | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| SR1948 | |
| Western Blot 1:500-1:2000 | |
| NR1F2nuclear receptor ROR-beta, Nuclear receptor RZR-beta, Nuclear receptor subfamily 1 group F member 2, RAR-related orphan receptor B, RAR-related orphan receptor beta, retinoic acid-binding receptor beta, retinoid-related orphan receptor beta, Retinoid-related orphan receptor-beta, ROR-BETA, RZRBbA133M9.1, RZR-BETA | |
| A synthesized peptide derived from human ROR beta (Uniprot #: Q92753) | |
| 100 μg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction | |
| 6096 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction