missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ ROR gamma Plasmid

Product Code. 18741674 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18741674 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18741674 Supplier R&D Systems™ Supplier No. NBP225278
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. For expression of hROR gamma protein in mammalian cells, induction of IL-17A promoter activity

The ROR gamma Plasmid is a mammalian expression vector construct, pCMV/hRORgamma, containing coding sequence of human RAR-related orphan receptor gamma.

  • Species: Human
  • Gene: RORC
  • Immunogen: Human ROR gamma Plasmid
  • Total contents of 10μg is enough for one 96-well cell transfection

Recommended Storage

Store at -20°C. Avoid freeze-thaw cycles.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Line Novus
Product Type ROR gamma Plasmid
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.