missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The ROR gamma (t) [del. 5-8] Plasmid is a mammalian expression vector construct, pCMV/hRORgamma (t) [del. 5-8], containing coding sequence of a novel isoform of human RAR-related orphan receptor gamma (t) which lacks the hingeencoding exons 5 - 8 and represses IL-17A gene transcription1.
- Species: Human
- Gene: RORC
- Immunogen: ROR gamma (t) [del. 5-8] Plasmid
- Total contents of 10μg is enough for one 96-well cell transfection
Recommended Storage
Store at -20°C. Avoid freeze-thaw cycles.
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Product Type | ROR gamma (t) [del. 5-8] Plasmid |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?