Learn More
Invitrogen™ ROR2 Recombinant Rabbit Monoclonal Antibody (24GB1065)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557239
 View more versions of this product
Product Code. 30276057
  • 500.00 € / 100µL
Description
ROR2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
ROR2 (receptor tyrosine kinase-like orphan receptor 2) is a type I transmembrane protein that belongs to the ROR subfamily of cell surface receptors. It is thought to be involved in the early formation of the chondrocytes and cartilage and growth plate development. This Type I membrane protein is expressed at high levels during early embryonic development. The expression levels drop strongly around day 16 and there are only very low levels in adult tissues. Defects in ROR2 are a cause of brachydactyly type B1 (BDB1). BDB1 is an autosomal dominant skeletal disorder characterized by hypoplasia/aplasia of distal phalanges and nails. In BDB1 the middle phalanges are short but in addition the terminal phalanges are rudimentary or absent. Both fingers and toes are affected. The thumbs and big toes are usually deformed. Defects in ROR2 are a cause of recessive Robinow syndrome (RRS). RRS is an autosomal disorder characterized by skeletal dysplasia with generalized limb bone shortening, segmental defects of the spine, brachydactyly and a dysmorphic facial appearance. The protein contains 1 frizzled (FZ) domain, 1 immunoglobulin-like C2-type domain, and 1 kringle domain.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ROR2
Recombinant Monoclonal
0.587 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q01974
ROR2
A synthesized peptide derived from human ROR2 (aa 30-82).
100 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1065
Unconjugated
ROR2
BDB; BDB1; mRor2; neurotrophic tyrosine kinase receptor-related 2; neurotrophic tyrosine kinase, receptor related 2; neurotrophic tyrosine kinase, receptor-related 2; NTRKR2; receptor tyrosine kinase like orphan receptor 2; receptor tyrosine kinase-like orphan receptor 2; ROR2; Tyrosine-protein kinase transmembrane receptor ROR2
Rabbit
Affinity chromatography
RUO
4920
-20°C