missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rosiglitazone, MedChemExpress
Click to view available options
Quantity:
50 mg
200 mg
Unit Size:
200mg
50mg
Description
Rosiglitazone (BRL 49653) is an orally active selective PPARγ agonist (EC50: 60 nM, Kd: 40 nM). Rosiglitazone is an TRPC5 activator (EC50: 30 μM) and TRPM3 inhibitor. Rosiglitazone can be used in the research of obesity and diabetes, senescence, ovarian cancer.
Specifications
Specifications
| Chemical Name or Material | Rosiglitazone |
| CAS | 122320-73-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H19N3O3S |
| Quantity | 200 mg |
| Synonym | BRL 49653 |
| Solubility Information | DMSO : 250 mg/mL (699.44 mM; Need ultrasonic) ∣Ethanol : 2 mg/mL (5.60 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble) |
| SMILES | O=C(N1)SC(CC2=CC=C(OCCN(C)C3=NC=CC=C3)C=C2)C1=O |
| Molecular Weight (g/mol) | 357.43 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction