missing translation for 'onlineSavingsMsg'
Learn More

Rosin, Gum, MP Biomedicals™

Product Code. 11410891 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410891 1 kg 1kg
11400891 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410891 Supplier MP Biomedicals Supplier No. 0215655791
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

acid value: 150 to 170

Chemical Identifiers

CAS 8050-09-7
Molecular Formula C20H30O2
Molecular Weight (g/mol) 302.46
MDL Number MFCD00132205
InChI Key RSWGJHLUYNHPMX-UHFFFAOYNA-N
Synonym 4bs,1r,10ar,4ar-1,4a-dimethyl-7-methylethyl-1,2,3,4,5,6,10,10a,4a,4b-decah ydrophenanthrenecarboxylic acid, 4ar-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid

Specifications

Color Orange
Density 1.07 - 1.09g/cm3 at 20°C
Quantity 1 kg
Physical Form Chunks
Chemical Name or Material Rosin, Gum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.