missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rosolic acid

Product Code. 10461461 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10461461 100 g Glass bottle 100g
10615102 25 g Glass bottle 25g
11976921 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10461461 Supplier Thermo Scientific Chemicals Supplier No. 229831000
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 603-45-2 | C19H14O3 | 290.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-45-2
Molecular Formula C19H14O3
Molecular Weight (g/mol) 290.32
MDL Number MFCD00001624
InChI Key FYEHYMARPSSOBO-UHFFFAOYSA-N
Synonym aurin, rosolic acid, corallin, p-rosolic acid, aurine, spirit aurine, corallin spirit soluble, 4,4'-dihydroxyfuchsone, aurin no. 555, 4-bis 4-hydroxyphenyl methylene cyclohexa-2,5-dienone
PubChem CID 5100
ChEBI CHEBI:34544
IUPAC Name 4-[bis(4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
SMILES OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)=C1C=CC(=O)C=C1

Specifications

Color Brown-Red
Loss on Drying 10% max. (110°C, 1 hr)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 g
Beilstein 08,361
Merck Index 15,872
Solubility Information Solubility in water: practically insoluble. Other solubilities: freely soluble in naoh,koh,70% h2so4 and hcl,freely soluble in alcohol,slightly soluble in ether and chloroform
Formula Weight 290.32
Physical Form Crystalline Powder
Chemical Name or Material Rosolic acid
Show More Show Less
compliance-icons
Product Identifier
  • Rosolic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.