missing translation for 'onlineSavingsMsg'
Learn More

Rosolic acid

Product Code. 11420347
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420347 100 g 100g
11410347 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420347 Supplier Thermo Scientific Alfa Aesar Supplier No. B21737.22
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 603-45-2 | C19H14O3 | 290.32 g/mol

Rosolic acid is used as a pH indicator in the range from 5.0 to 6.8. It acts as an intermediate in the manufacturing dyes. It is involved in the activation of heme oxygenase and in the inhibition of DNA fragmentation. It finds application in the selective isolation of coliform bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rosolic acid is used as a pH indicator in the range from 5.0 to 6.8. It acts as an intermediate in the manufacturing dyes. It is involved in the activation of heme oxygenase and in the inhibition of DNA fragmentation. It finds application in the selective isolation of coliform bacteria.

Solubility
Soluble in alcohol, sodium hydroxide, potassium hydroxide and strong acids. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-45-2
Molecular Formula C19H14O3
Molecular Weight (g/mol) 290.32
MDL Number MFCD00001624
InChI Key FYEHYMARPSSOBO-UHFFFAOYSA-N
Synonym aurin, rosolic acid, corallin, p-rosolic acid, aurine, spirit aurine, corallin spirit soluble, 4,4'-dihydroxyfuchsone, aurin no. 555, 4-bis 4-hydroxyphenyl methylene cyclohexa-2,5-dienone
PubChem CID 5100
ChEBI CHEBI:34544
IUPAC Name 4-[bis(4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
SMILES OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)=C1C=CC(=O)C=C1

Specifications

Quantity 100 g
Beilstein 2055205
Merck Index 14,881
Solubility Information Soluble in alcohol,sodium hydroxide,potassium hydroxide and strong acids. Insoluble in water.
Formula Weight 290.32
Chemical Name or Material Rosolic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.