missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rotatin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 624.00 €
Specifications
| Antigen | rotatin |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18429171
|
Novus Biologicals
NBP1-90555-25ul |
25 μL |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18203706
|
Novus Biologicals
NBP1-90555 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
rotatin Polyclonal specifically detects rotatin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| rotatin | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 25914 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TVVNILLDQSECSMVRREAAFILQNLLVIPMPTEIIKDYTWQGPCVHDEDSGLSLIGKPALQALLYHCHFYEHLNQMVKHCYLGRCMFDLNFSAFDRNSESNDLNGLDDSFKFW | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DKFZp434G145, FLJ26356, FLJ39085, rotatin | |
| RTTN | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title