Learn More
Scientific Industries™ Rotator Genie Standard Platform with Bands
Product Code. 11391484 Shop All Ohaus Products
Change view
Click to view available options
For Use With (Equipment):
Rotator Genie
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
11391484 Rotator Genie Each
1 options
Product Code. 11391484 Supplier Scientific Industries™ Supplier No. 83041458
  • ONLINE EXCLUSIVE PRICE
    175.35 €
    148.69 € / Pack of 1
    Save 26.66 €
    15% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Hold various types of vessels with this platform on the Rotator-Genie.
Standard platform with bands. Supplied with SI-210X. Can also be used on SI-220X.
TRUSTED_SUSTAINABILITY
Specifications
Model SI-2110
For Use With (Equipment) Rotator Genie