missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rotavirus Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB100-64396
This item is not returnable.
View return policy
Description
Rotavirus Polyclonal specifically detects Rotavirus in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Rotavirus | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2) with 0.09% Sodium Azide | |
| Sheep | |
| Protein G purified | |
| RUO | |
| NB100-64396 recognizes human rotavirus serotypes I, II, III, IV and VIII. Rotaviruses are RNA viruses from the family Reoviridae that are the the most common cause of gastroenteritis in infants. The nucleic acid is contained in a triple layered protein capsid consisting of outer capsid proteins, VP4 and VP7. Immunization was performed with live Rotavirus serotypes to avoid the risk of surface protein damage (VP7 and VP4). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 5 mg/mL | |
| ELISA 1:500-1:5000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Major inner capsid protein VP6, VP6 | |
| Five serotypes of group A Rotavirus strains Wa (serotype I), DS-1 (serotype II), Ito (III), HOCHI (IV) and 69M (VIII). | |
| 0.5 mL | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction