missing translation for 'onlineSavingsMsg'
Learn More

Rottlerin, 95+%

Product Code. 10337191 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mg
Packaging:
Glass bottle
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10337191 10 mg Glass bottle 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10337191 Supplier Thermo Scientific Supplier No. 328490100
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 82-08-6 | C30H28O8 | 516.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 82-08-6
Melting Point >200.0°C
Color Orange to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C30H28O8
MDL Number MFCD00017361
Quantity 10 mg
Merck Index 15, 8405
Synonym rottlerin, mallotoxin, kamalin, unii-e29lp3zmuh, e29lp3zmuh, chembl34241, bmk1-f10, e-1-6-3-acetyl-2,4,6-trihydroxy-5-methylphenyl methyl-5,7-dihydroxy-2,2-dimethyl-2h-1-benzopyran-8-yl-3-phenyl-2-propen-1-one, 1-6-3-acetyl-2,4,6-trihydroxy-5-methyl-benzyl-5,7-dihydroxy-2,2-dimethyl-2h-chromen-8-yl-3-phenyl-propenone, 1-6-3-acetyl-2,4,6-trihydroxy-5-methylphenyl methyl-5,7-dihydroxy-2,2-dimethyl-2h-1-benzopyran-8-yl-3-phenyl-2-propen-1-one
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmso,chloroform,ethanol
InChI Key DEZFNHCVIZBHBI-ZHACJKMWSA-N
SMILES CC1=C(C(=C(C(=C1O)C(=O)C)O)CC2=C(C3=C(C(=C2O)C(=O)C=CC4=CC=CC=C4)OC(C=C3)(C)C)O)O
IUPAC Name (E)-1-[6-[(3-acetyl-2,4,6-trihydroxy-5-methylphenyl)methyl]-5,7-dihydroxy-2,2-dimethylchromen-8-yl]-3-phenylprop-2-en-1-one
Molecular Weight (g/mol) 516.6
PubChem CID 5281847
ChEBI CHEBI:8899
Formula Weight 516.6
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Rottlerin
Show More Show Less
compliance-icons
Product Identifier
  • Rottlerin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.