Learn More
RPAIN Mouse, anti-Human, Clone: 2D8E3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68546-1-IG-20UL
 View more versions of this product
Product Code. 18015155
  • 145.00 € / 20µL
Description
The RPAIN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RPAIN. This antibody recognizes human antigen. The RPAIN antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
RPAIN
2D8E3
HRIP, RIP, RPA interacting protein, RPAIN
20 μL
84268
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human
150 kDa
IgG1