Learn More
RPAIN Mouse, anti-Human, Clone: 2D8E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68546-1-IG-150UL
This item is not returnable.
View return policy
Description
The RPAIN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RPAIN. This antibody recognizes human antigen. The RPAIN antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| RPAIN | |
| 2D8E3 | |
| HRIP, RIP, RPA interacting protein, RPAIN | |
| 150 μL | |
| 84268 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |