missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL10A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00 € - 481.00 €
Specifications
| Antigen | RPL10A |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18683241
|
Novus Biologicals
NBP2-93591-0.02ml |
0.02 mL |
204.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18612431
|
Novus Biologicals
NBP2-93591-0.1ml |
0.1 mL |
481.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPL10A Polyclonal antibody specifically detects RPL10A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| RPL10A | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 4736 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CSA-19, Csa-19,60S ribosomal protein L10a, NEDD6, NEDD-6, Neural precursor cell expressed developmentally down-regulated protein 6, ribosomal protein L10a | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human RPL10A (NP_009035.3). MSSKVSRDTLYEAVREVLHGNQRKRRKFLETVELQISLKNYDPQKDKRFSGTVRLKSTPRPKFSVCVLGDQQHCDEAKAVDIPHMDIEALKKLNKNKKLVKKLAKKYDAF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title