missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL10A Antibody (OTI2G9), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
256.00 € - 550.00 €
Specifications
| Antigen | RPL10A |
|---|---|
| Clone | OTI2G9 |
| Dilution | Western Blot 1:500-2000, Immunohistochemistry 1:2000 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306477
|
Novus Biologicals
NBP3-44967-100ul |
100 μL |
550.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306368
|
Novus Biologicals
NBP3-44967-30ul |
30 μL |
256.00 €
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPL10A Monoclonal antibody specifically detects RPL10A in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| RPL10A | |
| Western Blot 1:500-2000, Immunohistochemistry 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CSA-19, Csa-19,60S ribosomal protein L10a, NEDD6, NEDD-6, Neural precursor cell expressed developmentally down-regulated protein 6, ribosomal protein L10a | |
| Full length human recombinant protein of human RPL10A (NP_009035) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI2G9 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 4736 | |
| IgG2a | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title