missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL13 Antibody (SR1422), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22510-50ul
This item is not returnable.
View return policy
Description
RPL13 Monoclonal antibody specifically detects RPL13 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| RPL13 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| SR1422 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| BBC1FLJ27454, Breast basic conserved protein 1, D16S444E, FLJ27453, MGC117342, MGC71373,60S ribosomal protein L13, OK/SW-cl.46, ribosomal protein L13 | |
| A synthesized peptide derived from human RPL13 (Uniprot #: P26373) | |
| 50 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6137 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction