missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPL13A Monoclonal Antibody (4C9)
GREENER_CHOICE

Product Code. 17931861
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. Quantity unitSize
17931861 100 μg 100µg
1 options
This item is not returnable. View return policy

Product Code. 17931861

Brand: Invitrogen™ MA549258

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hek293 whole cell, human Hela whole cell, human K562 whole cell. ICC/IF: CACO-2 cell. Flow: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L13P family of ribosomal proteins. It is located in the cytoplasm. Transcript variants utilizing alternative polyA signals have been observed. This gene is co-transcribed with the small nucleolar RNA genes U32, U33, U34, and U35, which are located in its second, fourth, fifth, and sixth introns, respectively. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL13A
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4C9
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene RPL13A
Gene Accession No. P40429
Gene Alias 1810026N22Rik; 23 kDa highly basic protein; 60S ribosomal protein L13a; L13A; Large ribosomal subunit protein uL13; P198; ribosomal protein L13a; Rpl13a; tissue specific transplantation antigen 1; tissue specific transplantation antigen P198-7; transplantation antigen P198; TSTA1; Tstap198-7; tum-antigen; Tum-P198 antigen; tum-transplantation antigen P198
Gene Symbols RPL13A
Host Species Mouse
Immunogen E.coli-derived human RPL13A recombinant protein (Position:A2-V203).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23521
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.