missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL7A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13258-25ul
This item is not returnable.
View return policy
Description
RPL7A Polyclonal antibody specifically detects RPL7A in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| RPL7A | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
| 60S ribosomal protein L7a, PLA-X polypeptide, ribosomal protein L7a, SURF-3, SURF3surfeit 3, Surfeit locus protein 3, thyroid hormone receptor uncoupling protein, TRUP | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: EAIRTNYNDRYDEIRRHWGGNVLGPKSVARIAKLE | |
| 25 μL | |
| Apoptosis | |
| 6130 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunocytochemistry | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur