missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL7A Antibody (SR1517), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21877-100ul
This item is not returnable.
View return policy
Description
RPL7A Monoclonal antibody specifically detects RPL7A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| RPL7A | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1517 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200 | |
| 60S ribosomal protein L7a, PLA-X polypeptide, ribosomal protein L7a, SURF-3, SURF3surfeit 3, Surfeit locus protein 3, thyroid hormone receptor uncoupling protein, TRUP | |
| A synthesized peptide derived from human RPL7A (Uniprot #: P62424) | |
| 100 μg | |
| Apoptosis | |
| 6130 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction