missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPLP0 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
230.00 € - 317.00 €
Specifications
| Antigen | RPLP0 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:100-1:500 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18670982
|
Novus Biologicals
NBP2-97776-100ul |
100 μg |
317.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603305
|
Novus Biologicals
NBP2-97776-50ul |
230.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
RPLP0 Polyclonal antibody specifically detects RPLP0 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| RPLP0 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| 60S acidic ribosomal protein P0, 60S ribosomal protein L10E, acidic ribosomal phosphoprotein P0, L10E, MGC111226, MGC88175, P0, PRLP0, ribosomal protein, large, P0, RPP0 | |
| Produced in rabbits immunized with E. coli-derived Human RPLP0 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 6175 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title