missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rpmB/L28 Antibody - N-Terminus - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
199.00 € - 462.00 €
Specifications
| Antigen | rpmB/L28 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5 ug/ml, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682055
|
Novus Biologicals
NBP3-23514-0.025mg |
0.025 mg |
199.00 €
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647504
|
Novus Biologicals
NBP3-23514-0.1mg |
0.1 mg |
462.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
rpmB/L28 Polyclonal antibody specifically detects rpmB/L28 in E. coli samples. It is validated for Western Blot, ELISASpecifications
| rpmB/L28 | |
| Western Blot 0.5 ug/ml, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS | |
| 946941 | |
| IgG | |
| Affinity purified |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| E. coli | |
| 50S ribosomal subunit protein L28, ECK3627, Large ribosomal subunit protein bL28, rpmB | |
| Antibody was raised against an 15 amino acid peptide near the amino terminus of E. coli rpmB/L28. The immunogen is located within the first 50 amino acids of rpmB/L28. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title