missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ RPMI 1640 Medium, HEPES
GREENER_CHOICE

Product Code. 12017599
Change view
Click to view available options
Quantity:
10 x 500 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12017599 10 x 500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12017599 Supplier Gibco™ Supplier No. 52400041

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. With L-Glutamine, 25 mM HEPES.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mammalian Cells
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type RPMI 1640
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
With Additives Glutamine, HEPES, Phenol Red, Sodium Bicarbonate
Without Additives No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 10 x 500 mL
Osmolality 270 - 310 mOsm/kg
pH 7 to 7.4
Content And Storage Storage conditions: 2-8° C. Protect from light.
Shipping conditions: Ambient.
Shelf life: 12 months from date of manufacture.
Culture Environment CO2
Culture Type Mammalian Cell Culture
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.