missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ RPMI 1640 Medium, powder

Product Code. 10655953 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 L
1 L
50 L
Unit Size:
1 L
10 L
50 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10655953 1 L 1 L
11544506 10 L 10 L
11554506 50 L 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10655953 Supplier Gibco™ Supplier No. 51800019
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

RPMI 1640 Medium, powder

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 has since been found suitable for a variety of mammalian cells including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. Life Technologies offers a variety of Gibco™ RPMI 1640 modifications for a range of cell culture applications.

Gibco™ RPMI 1640 is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 contains biotin, vitamin B12, and PABA which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. Customers using Gibco™ RPMI 1640 in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from Life Technologies to reference our Type II Drug Master File (DMF).

cGMP Manufacturing and Quality System
Gibco™ RPMI 1640 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, Life Technologies offers a comparable Gibco™ RPMI 1640 product made in our Grand Island facility (31800-089). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

RPMI 1640 contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Powder forms of Gibco™ L-15 cell culture medium require sodium bicarbonate supplementation pH adjustment, and filtration at the time of preparation.
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Roswell Park Memorial Institute (RPMI) 1640 Medium
With Additives Glutamine, Phenol Red
Without Additives No HEPES, No Sodium Pyruvate, No Sodium Bicarbonate
For Use With (Application) Mammalian cell culture
Quantity 1 L
Certifications/Compliance ISO 13485 standards, FDA
pH 7.4 to 7.6
Content And Storage 2 to 8°C
Description Roswell Park Memorial Institute (RPMI) 1640 Medium, 1L, Powder, 7.4 to 7.6 pH, With L-Glutamine, Phenol Red, 36 Months Shelf Life, 230 to 280mOsm/kg Osmolality, For Mammalian Cell Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.