missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein S10 (RPS10) is a 165 amino acid component of the small ribosomal 40S subunit. While generally found in the cytoplasm, RPS10 can localize to the granular component (GC) region of the nucleolus. For this to occur, methylation by PRMT5 is required as well as an interaction with NPS1. Defects in RPS10 are the cause of Diamond-Blackfan anemia type 9 (DBA9). It is a form of Diamond-Blackfan anemia, a congenital non-regenerative hypoplastic anemia that usually presents early in infancy. Diamond-Blackfan anemia is characterized by a moderate to severe macrocytic anemia, erythroblastopenia, and an increased risk of maligcy.
Specifications
Specifications
| Antigen | RPS10 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.17 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RPS10 |
| Gene Accession No. | P46783, P63325, P63326 |
| Gene Alias | 40S ribosomal protein S10, ribosomal protein S10, RPS10 |
| Gene Symbols | RPS10 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?