Learn More
Invitrogen™ RPS6KB2 Recombinant Rabbit Monoclonal Antibody (24GB5075)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557694
 View more versions of this product
Product Code. 30276265
  • 564.00 € / 100µL
Description
RPS6KB2 Recombinant Monoclonal Antibody for Western Blot, Flow
RPS6KB2 (p70S6K beta) is a serine/threonine kinase with 2 non-identical kinase catalytic domains which phosphorylates the S6 ribosomal protein and eukaryotic translation initiation factor 4B (eIF4B). Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RPS6KB2
Recombinant Monoclonal
Unconjugated
Rps6kb2
70 kDa ribosomal protein S6 kinase 2; 70kDa; KLS; p70 ribosomal S6 kinase beta; p70 S6 kinase beta; p70 S6KB; p70 S6K-beta; p70(S6K)-beta; P70-beta; P70-beta-1; P70-beta-2; p70-S6K 2; P70S6K2; p70S6Kb; ribosomal protein S6 kinase B2; ribosomal protein S6 kinase beta-2; ribosomal protein S6 kinase, 70kD, polypeptide 2; ribosomal protein S6 kinase, 70kDa, polypeptide 2; ribosomal protein S6 kinase, polypeptide 2; RPS6KB2; S6 kinase 2; S6 kinase-related kinase; S6K2; S6K-beta; S6K-beta2; S6K-beta-2; serine/threonine-protein kinase 14 beta; serine/threonine-protein kinase 14B; SRK; STK14B
Rabbit
Affinity chromatography
RUO
361696, 58988, 6199
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB5075
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9UBS0, Q9Z1M4
Rps6kb2
Recombinant protein of human P70 S6 Kinase beta (aa 1-120).
100 μL
Primary
Human, Mouse, Rat