missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPUSD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88437
This item is not returnable.
View return policy
Description
RPUSD2 Polyclonal antibody specifically detects RPUSD2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RPUSD2 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| C15orf19, C18B11, C18B11 homolog (44.9kD), chromosome 15 open reading frame 19, FLJ31409, RNA pseudouridylate synthase domain containing 2, RNA pseudouridylate synthase domain-containing protein 2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: VFSTEFRAQPLAYYEAAVRAGRLQLNEKPVQDLNIVLKDNDFLRNTVHRHEPPVTAEPIRLLAENEDVVVVDKPSSIPVHPCGRFRHNTVIFILGKEHQLKELH | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 27079 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction