missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRM2 Antibody (SAIC-30C-18) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20098-0.2mg
This item is not returnable.
View return policy
Description
RRM2 Monoclonal antibody specifically detects RRM2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Mass Spectrometry
Specifications
| RRM2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Mass Spectrometry | |
| SAIC-30C-18 | |
| Unconjugated | |
| EC 1.17.4.1, R2, ribonucleoside-diphosphate reductase subunit M2, ribonucleotide reductase M2, ribonucleotide reductase M2 polypeptide, Ribonucleotide reductase small chain, Ribonucleotide reductase small subunit, RR2, RR2M | |
| The KLH-conjugated peptide 'IEQEFLTEALPVK' derived from Ribonucleoside-diphosphate reductase subunit M2 (RRM2). RRM2 provides precursors required for DNA synthesis. | |
| 0.2 mg | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 6241 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction