Learn More
RRM2 Mouse, anti-Human, CoraLite Plus 488, Clone: 2A9A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67006-20UL
Description
The RRM2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RRM2. This antibody recognizes human antigen. The RRM2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| RRM2 | |
| 2A9A7 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| R2, RR2, RR2M, RRM2 | |
| 20 μL | |
| 6241 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |