missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RS 17053 hydrochloride, MedChemExpress
Description
RS 17053 hydrochloride is a potent and selective α1A adrenoceptor antagonist, with a pKi value of 9.1 in native cell membrane and a pA2 value of 9.8 in functional assays.
Specifications
Specifications
| Chemical Name or Material | RS 17053 hydrochloride |
| CAS | 169505-93-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H30Cl2N2O2 |
| Quantity | 50 mg |
| Synonym | RS-17053 |
| Solubility Information | DMSO : ≥ 125 mg/mL (278.14 mM) |
| SMILES | [H]Cl.ClC1=CC2=C(NC=C2CC(C)(C)NCCOC3=CC=CC=C3OCC4CC4)C=C1 |
| Molecular Weight (g/mol) | 449.41 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?