missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (RS)-CPP
GREENER_CHOICE

Product Code. 18064882
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18064882 10 mg 10mg
30001420 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18064882 Supplier Tocris Bioscience™ Supplier No. 0173/10
  • ONLINE EXCLUSIVE PRICE
    280.00 €
    264.60 € / 10mg
    Save 15.40 €
    5% Off
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent NMDA antagonist

Potent NMDA antagonist.

R-enantiomer also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100828-16-8
Molecular Formula C8H17N2O5P

Specifications

Quantity 10 mg
Formula Weight 252.21
Purity >98%
Molecular Weight (g/mol) 252.207
Chemical Name or Material (RS)-CPP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.