Learn More
RSAD2 Mouse, anti-Human, Clone: 1C5D4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68604-1-IG-20UL
 View more versions of this product
Product Code. 18020626
  • 145.00 € / 20µL
Description
The RSAD2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RSAD2. This antibody recognizes human, mouse, pig antigen. The RSAD2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
RSAD2
1C5D4
2510004L01Rik, cig33, cig5, RSAD2, vig1, Viperin
20 μL
91543
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse, Pig
150 kDa
IgG1