Learn More
RSAD2 Mouse, anti-Human, Clone: 1C5D4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68604-1-IG-150UL
This item is not returnable.
View return policy
Description
The RSAD2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RSAD2. This antibody recognizes human, mouse, pig antigen. The RSAD2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| RSAD2 | |
| 1C5D4 | |
| 2510004L01Rik, cig33, cig5, RSAD2, vig1, Viperin | |
| 150 μL | |
| 91543 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Pig | |
| 150 kDa | |
| IgG1 |