missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSK1 Recombinant Rabbit Monoclonal Antibody (23GB2080), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
207.00 € - 562.00 €
Specifications
| Antigen | RSK1 |
|---|---|
| Clone | 23GB2080 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088530
![]() |
Invitrogen™
MA553216 |
100 μL |
562.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088212
|
Invitrogen™
MA553217 |
20 μL |
207.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RSK1 Recombinant Monoclonal antibody specifically detects RSK1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| RSK1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q15418 | |
| 6195 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2080 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 90 kDa ribosomal protein S6 kinase 1; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); hu-1; MAP kinase-activated protein kinase 1; MAP kinase-activated protein kinase 1a; MAPK-activated protein kinase 1; MAPK-activated protein kinase 1a; MAPKAP kinase 1; MAPKAP kinase 1a; MAPKAPK-1; mapkapk1a; MAPKAPK-1a; p90Rsk; P90-RSK; p90-RSK 1; p90Rsk1; p90-Rsk1; p90S6K; pp90RSK1; ribosomal protein S6 kinase 2 alpha; ribosomal protein S6 kinase 2 beta; ribosomal protein S6 kinase a, polypeptide 1; ribosomal protein S6 kinase A1; ribosomal protein S6 kinase A1 L homeolog; ribosomal protein S6 kinase alpha 1; Ribosomal protein S6 kinase alpha-1; Ribosomal protein S6 kinase II alpha; ribosomal protein S6 kinase II beta; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 90kD, polypeptide 1; Ribosomal protein S6 kinase, 90kD, 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1 L homeolog; ribosomal protein S6 kinase, polypeptide A1; ribosomal S6 kinase 1; RPS6KA; Rps6ka1; rps6ka1.L; RPS6KA1L; rsk; RSK1; Rsk-1; S6 kinase II beta; S6 protein kinase (Rsk-1); S6K-alpha 1; S6K-alpha-1; S6KII-alpha; S6KII-beta; wu:fc56h07; XELAEV_18012109mg; zgc:152654 | |
| A synthesized peptide derived from human RSK1 p90 (1-67AA). | |
| Primary | |
| -20°C | |
| RPS6KA1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title