missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSK3 Antibody (S05-5E4), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00 € - 645.00 €
Specifications
| Antigen | RSK3 |
|---|---|
| Clone | S05-5E4 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 1:20-1:50, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18696663
|
Novus Biologicals
NBP3-19811-100ul |
100 μg |
645.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656353
|
Novus Biologicals
NBP3-19811-50ul |
50 μg |
421.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RSK3 Monoclonal antibody specifically detects RSK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| RSK3 | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 6196 | |
| IgG | |
| Affinity purified |
| S05-5E4 | |
| Western Blot, Immunohistochemistry 1:20-1:50, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.11, EC 2.7.11.1, HU-2, MAP kinase-activated protein kinase 1c, MAPK-activated protein kinase 1c, MAPKAP kinase 1c, MAPKAPK-1c, MAPKAPK1C, p90-RSK 2, p90RSK2, p90-RSK3, ribosomal protein S6 kinase alpha 2, ribosomal protein S6 kinase alpha-2, ribosomal protein S6 kinase, 90kD, polypeptide 2, ribosomal protein S6 kinase, 90kDa, polypeptide 2, Ribosomal S6 kinase 3,90 kDa ribosomal protein S6 kinase 2, RSK, RSK-3, RSK3pp90RSK3, S6K-alpha, S6K-alpha2, S6K-alpha-2 | |
| A synthetic peptide of human RSK3 (Uniprot # Q15349) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title