missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSV/BRSV Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00 € - 458.00 €
Specifications
| Antigen | RSV/BRSV |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18292951
|
Novus Biologicals
NB100-93586SS |
0.025 mg |
168.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18717513
|
Novus Biologicals
NB100-93586 |
458.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
RSV/BRSV Polyclonal specifically detects RSV/BRSV in Virus samples. It is validated for Western Blot.Specifications
| RSV/BRSV | |
| Western Blot 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Virus | |
| Bovine Respiratory Syncytial Virus BRSV, Bovine RSV | |
| IgG | |
| Affinity Purified | |
| Reacts with Human Respiratory Syncytial Virus (RSV) and Bovine Respiratory Syncytial Virus (BRSV) Fusion protein of all strains of RSV BRSV in human and bovine samples. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Residues 483-488 [FPSDEFC] of the 63 kDa RSV and BRSV F protein. This sequence represents a neutralizing epitope and is 100% conserved among all known species. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title