missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RTCD1 catalyzes the conversion of 3′-phosphate to a 2′, 3′-cyclic phosphodiester at the end of RNA. The mechanism of action of the enzyme occurs in 3 steps: (A) adenylation of the enzyme by ATP; (B) the enzyme acts on RNA-N3′P to produce RNA-N3′PP5′A; (C) a non catalytic nucleophilic attack by the adjacent 2′hydroxyl on the phosphorus in the diester linkage to produce the cyclic end product. The biological role of this enzyme is unknown but it is likely to function in some aspects of cellular RNA processing. There are two named isoforms.
Specifications
Specifications
| Antigen | RTCD1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RTCA |
| Gene Accession No. | O00442, Q9D7H3 |
| Gene Alias | RNA 3 phosphate cyclase, RNA cyclase, RPC, RPC1, RTC1, RTCD1 |
| Gene Symbols | RTCA, Rtcd1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?