missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTN1-A/NSP Antibody (RNL-4), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97673MFV500
This item is not returnable.
View return policy
Description
RTN1-A/NSP Monoclonal antibody specifically detects RTN1-A/NSP in Human, Rat, Porcine samples. It is validated for Flow Cytometry
Specifications
| RTN1-A/NSP | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Pig | |
| Purified |
| Flow Cytometry | |
| RNL-4 | |
| Flow Cytometry | |
| NSP | |
| Derived by fusion of SP2/0-Ag14 Mouse myeloma cells, with spleen cells from a BALB/c Mouse immunized with a synthetic peptide encompassing the unique 20 N-terminal amino acid sequence of Reticulon-1C. | |
| 0.1 mL | |
| Neuroscience | |
| 6252 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction