missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTVP-1/GLIPR1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94550-0.02ml
This item is not returnable.
View return policy
Description
RTVP-1/GLIPR1 Polyclonal antibody specifically detects RTVP-1/GLIPR1 in Rat samples. It is validated for Western Blot
Specifications
| RTVP-1/GLIPR1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| GLI pathogenesis-related 1, GLI pathogenesis-related 1 (glioma), GliPR, GliPR 1, GLIPRglioma pathogenesis-related protein 1, Protein RTVP-1, related to testis-specific, vespid, and pathogenesis proteins 1, RTVP1CRISP7, testes-specific vespid and pathogenesis protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 22-140 of human GLIPR1 (NP_006842.2). ANILPDIENEDFIKDCVRIHNKFRSEVKPTASDMLYMTWDPALAQIAKAWASNCQFSHNTRLKPPHKLHPNFTSLGENIWTGSVPIFSVSSAITNWYDEIQDYDFKTRICKKVCGHYTQ | |
| 0.02 mL | |
| Apoptosis, Cancer, Cell Biology | |
| 11010 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction