missing translation for 'onlineSavingsMsg'
Learn More

RU.521, MedChemExpress

Product Code. 16487537
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487537

Brand: Medchem Express HY114180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RU.521 (RU320521) is a potent and selective cyclic GMP-AMP synthase (cGAS) inhibitor and inhibits cGAS-mediated interferon upregulation. RU.521 suppresses dsDNA-activated reporter activity with an IC50 of 0.7 μM. RU.521 reduces constitutive expression of interferon in macrophages from a mouse model of Aicardi-Goutières syndrome (AGS).

RU.521 (RU320521) is a potent and selective cyclic GMP-AMP synthase (cGAS) inhibitor and inhibits cGAS-mediated interferon upregulation. RU.521 suppresses dsDNA-activated reporter activity with an IC50 of 0.7 μM. RU.521 reduces constitutive expression of interferon in macrophages from a mouse model of Aicardi-Goutières syndrome (AGS).

Specifications

Chemical Name or Material RU.521
Physical Form Solid
CAS 2262452-06-0
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H12Cl2N4O3
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym RU320521
Solubility Information DMSO : 83.33 mg/mL (200.68 mM; Need ultrasonic)
SMILES O=C1OC(C2=C(O)N(C3=NC4=CC=C(Cl)C(Cl)=C4N3)N=C2C)C5=C1C=CC=C5
Molecular Weight (g/mol) 415.23
Formula Weight 415.23
Percent Purity 98.67%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.