missing translation for 'onlineSavingsMsg'
Learn More

Rubrene, 99%

Product Code. 10346970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10346970 500 mg Glass bottle 500mg
10449060 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10346970 Supplier Thermo Scientific Acros Supplier No. 132385000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 517-51-1 | C42H28 | 532.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 517-51-1
Molecular Formula C42H28
Molecular Weight (g/mol) 532.67
MDL Number MFCD00003703
InChI Key YYMBJDOZVAITBP-UHFFFAOYSA-N
Synonym rubrene, 5,6,11,12-tetraphenylnaphthacene, rubren, naphthacene, 5,6,11,12-tetraphenyl, rubrene, powder, acmc-209kvj, 5,11,12-tetraphenyltetracene, 5,11,12-tetraphenylnaphthacene, naphthacene,6,11,12-tetraphenyl
PubChem CID 68203
IUPAC Name 5,6,11,12-tetraphenyltetracene
SMILES C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8

Specifications

Melting Point 330°C to 335°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Beilstein 05, II, 726
Solubility Information Solubility in water: practically insoluble
Formula Weight 532.67
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Rubrene, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.