Learn More
Ruthenium(III) chloride hydrate, Premion™, 99.99% (metals basis)
Description
Ruthenium(III) chloride hydrate is used as a starting material in ruthenium chemistry. It acts as a precursor to many ruthenium compounds, which finds application as catalysts in asymmetric hydrogenation and alkene metathesis. It serves as a catalyst in the preparation of 2-ethyl-3-methylquinolines from primary aromatic amines and triallylamine. Further, it is used in the preparation of nanoparticulate ruthenium-aluminum oxyhydroxide catalyst for the efficient dehydrogenation of arylmethyl alcohols to the corresponding aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 100°C (decomposition) |
| Color | Black |
| Physical Form | Crystalline |
| Quantity | 10 g |
| UN Number | UN3260 |
| Merck Index | 14,8302 |
| Solubility Information | Soluble in water. |
| Formula Weight | 207.43 (Anhydrous) |
| Percent Purity | 99.99% |
| Grade | Premion |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.